Glassy Carbon Crucibles for Crystal Growth, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Glassy carbon offers high purity, corrosion resistance, thermal stability, and a structure impermeable to both gases and liquids
- Resistance to all wet decomposition agents
- No memory effects (uncontrolled adsorption and release of foreign elements)
- No contamination of analytical samples
- Stability to acid and alkaline melts
- No wetting effect of metal melts
- Good resistance to thermal shock allows rapid heating and cooling times
- Resistive to abrasive wear
- Good electrical conductivity
These crucibles are designed for high-temperature differential thermal/thermal gravimetric analyses and ultra-high purity materials technology involving crystal growing.
Alta Aesar's glassy carbon products are subjected to a high temperature heat treatment which imparts special material properties, including significantly improved corrosion resistance and strength.