Order Entry
United States
ContactUsLinkComponent
Phosphorus, Plasma Standard Solution, Specpure®, P 1000 µg/ml, Thermo Scientific
Phosphorus, Plasma Standard Solution, Specpure®, P 1000 µg/ml, Thermo Scientific
Catalog # AA13862-AP
CAS Number:  
Phosphorus, Plasma Standard Solution, Specpure®, P 1000 µg/ml, Thermo Scientific
Catalog # AA13862-AP
Supplier Number:  013862-AP
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    5% HNO₃
  • Size:
    500 ml
  • MDL No.:
    MFCD00133771
  • Cat. No.:
    AA13862-AP
  • Accuracy:
    ±1%
  • Element:
    Phosphorus
  • Supplier no.:
    13862-AP

Specifications

About this item

Molecular Formula: P in 5% HNO3
Physical Form: Liquid
Appearance: Colorless
UN No.: 3264
MDL No.: MFCD00133771