ContactUsLinkComponent
Sklar Black™ Collin Vaginal Speculum, OR Grade, Sklar
Sklar Black™ Collin Vaginal Speculum, OR Grade, Sklar
  10197-948
 :  Sklar
undefined
Sklar Black™ Collin Vaginal Speculum, OR Grade, Sklar
  10197-948
 :  Sklar
 :  90-3946
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Sklar Black™ Collin Vaginal Speculum
  • Material:
    Stainless Steel with Reduced Reflectivity Polymer Coating
  • Size:
    Medium
  • Cat. No.:
    10197-948
  • Supplier no.:
    90-3946
The Sklar Black™ Collin Vaginal Speculum is a gynecological instrument.
  • Used for performing pelvic examinations
  • Pap Smears
  • Cervix exams
It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. Unlike many other vaginal specula, there is no handle with the Collin device and the blades open laterally/horizontally versus vertically.
The practitioner can best determine which type and size speculum is most appropriate for the patient. This product features a permanent smoke evacuation tube (not shown).
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries.