ContactUsLinkComponent
Corrugated Sample Storage Tray
Corrugated Sample Storage Tray
Catalog # 10754-106
undefined
Corrugated Sample Storage Tray
Catalog # 10754-106
Supplier Number:  HS120375
Specifications
  • Capacity:
    24 full specimen containers
  • Color:
    White
  • Description:
    Corrugated sample storage tray
  • Dimensions:
    40.6L×26W×8.9H cm (16×10.2×3.5")
  • Cat. No.:
    10754-106
  • Supplier no.:
    HS120375
Specifications
About this item
Corrugated storage trays can be used with the included dividers for specimen containers or remove them for general lab product storage or transport. Affordable sample organization and storage.
  • Made from corrugated polypropylene
  • Moisture resistant, significantly reducing the chance of mold
  • Lightweight and sturdy to hold up to 24 full specimen containers
  • Stackable with other trays
  • Removable dividers allow outer shell to be used for storage or transport of laboratory products
Ordering information: Supplied flat-packed, each order includes 10 trays and 10 dividers.