ContactUsLinkComponent
Spatula and Packer, OR Grade, Sklar
Spatula and Packer, OR Grade, Sklar
Catalog # 10192-426
Supplier:  Sklar
undefined
Spatula and Packer, OR Grade, Sklar
Catalog # 10192-426
Supplier:  Sklar
Supplier Number:  97-0542
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Spatula and Packer
  • Length:
    5¾"
  • Cat. No.:
    10192-426
  • Supplier no.:
    97-0542
Specifications
About this item
Sklar's Spatula and Packer is double-ended instrument commonly used in podiatry.
  • It features one smooth, rounded teardrop tip (spatula) and one blunt probe end (packer)
  • This product is straight and 53/4" long
The spatula end is used to separate the nail plate from the nail bed. The packer is used to place packing under or around the toe following a surgical procedure.