Order Entry
United States
ContactUsLinkComponent
 
Anti-IL9 Rat Monoclonal Antibody [clone: 222604]
  220020-741
Anti-IL9 Rat Monoclonal Antibody [clone: 222604]
  220020-741
 :  MAB409

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Interleukin 9
  • Antigen Symbol:
    IL9
  • Clonality:
    Monoclonal
  • Clone:
    222604
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    500 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    16198
  • Antigen Synonyms:
    Interleukin-9|IL-9|IL-9 |Cytokine P40|HP40|P40
  • Amino Acid Number:
    Gln19 to Pro144
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL‑9 Gln19-Pro144; Accession # P15247.1
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220020-741
  • Supplier no.:
    MAB409

 

 

Type: Primary
Antigen: IL9
Clonality: Monoclonal
Clone: 222604
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse