Order Entry
United States
ContactUsLinkComponent
Anti-CCL27 Mouse Monoclonal Antibody [clone: 124302]
Anti-CCL27 Mouse Monoclonal Antibody [clone: 124302]
Catalog # 220020-605
Supplier:  R&D Systems
Anti-CCL27 Mouse Monoclonal Antibody [clone: 124302]
Catalog # 220020-605
Supplier:  R&D Systems
Supplier Number:  MAB3761

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Chemokine (C-C motif) ligand 27
  • Antigen Symbol:
    CCL27
  • Clonality:
    Monoclonal
  • Clone:
    124302
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    500 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    10850
  • Antigen Synonyms:
    PESKY|ILC|ESKINE|CTACK|SCYA27|CTAK|ALP
  • Amino Acid Number:
    Phe25 to Gly112
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    E. coli -derived recombinant human CCL27/CTACK Phe25-Gly112; Accession # NP_006655
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220020-605
  • Supplier no.:
    MAB3761

Specifications

About this item

Type: Primary
Antigen: CCL27
Clonality: Monoclonal
Clone: 124302
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human