Order Entry
United States
ContactUsLinkComponent
Anti-RGMB Mouse Monoclonal Antibody [clone: 398528]
Anti-RGMB Mouse Monoclonal Antibody [clone: 398528]
  220020-548
Anti-RGMB Mouse Monoclonal Antibody [clone: 398528]
  220020-548
 :  MAB3630

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Repulsive Guidance Molecule B
  • Antigen Symbol:
    RGMB
  • Clonality:
    Monoclonal
  • Clone:
    398528
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    285704
  • Antigen Synonyms:
    Rgmb|Dragon|RGM b|RGM domain family member B|RGMB_HUMAN.|Repulsive Guidance Molecule B|DRG11-responsive axonal guidance and outgrowth of neurite
  • Amino Acid Number:
    Gly46 to Ser411
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant human RGM-B isoform 1 Gly46-Ser411; Accession # Q6NW40
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220020-548
  • Supplier no.:
    MAB3630

 

 

Type: Primary
Antigen: RGMB
Clonality: Monoclonal
Clone: 398528
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human