Order Entry
United States
ContactUsLinkComponent
 
Anti-CDH6 Mouse Monoclonal Antibody [clone: 427903]
  220020-050
Anti-CDH6 Mouse Monoclonal Antibody [clone: 427903]
  220020-050
 :  MAB27151

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Cadherin 6
  • Antigen Symbol:
    CDH6
  • Clonality:
    Monoclonal
  • Clone:
    427903
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG3
  • Reactivity:
    Human
  • Size:
    100 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in Tris and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    1004
  • Antigen Synonyms:
    KCAD|CAD6|K-Cadherin (fetal kidney)
  • Amino Acid Number:
    Ser54 to Ala615
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant human Cadherin‑6/KCAD Ser54-Ala615; Accession # P55285
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220020-050
  • Supplier no.:
    MAB27151

 

 

Type: Primary
Antigen: CDH6
Clonality: Monoclonal
Clone: 427903
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: