Order Entry
United States
ContactUsLinkComponent
Anti-PTPN9 Mouse Monoclonal Antibody [clone: 291835]
Anti-PTPN9 Mouse Monoclonal Antibody [clone: 291835]
Catalog # 220020-020
Supplier:  R&D Systems
Anti-PTPN9 Mouse Monoclonal Antibody [clone: 291835]
Catalog # 220020-020
Supplier:  R&D Systems
Supplier Number:  MAB2668

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Protein Tyrosine Phosphatase, Non-receptor Type 9
  • Antigen Symbol:
    PTPN9
  • Clonality:
    Monoclonal
  • Clone:
    291835
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    56294
  • Antigen Synonyms:
    MEG2
  • Amino Acid Number:
    Glu2 to Gln593
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    E. coli -derived recombinant human PTP-MEG2 Glu2-Gln593; Accession # P43378
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220020-020
  • Supplier no.:
    MAB2668

Specifications

About this item

Type: Primary
Antigen: PTPN9
Clonality: Monoclonal
Clone: 291835
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat