Order Entry
United States
ContactUsLinkComponent
Anti-ITGAV Mouse Monoclonal Antibody [clone: 273210]
Anti-ITGAV Mouse Monoclonal Antibody [clone: 273210]
Catalog # 220018-910
Supplier:  R&D Systems
Anti-ITGAV Mouse Monoclonal Antibody [clone: 273210]
Catalog # 220018-910
Supplier:  R&D Systems
Supplier Number:  MAB12191

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Integrin Alpha 5
  • Antigen Symbol:
    ITGA5
  • Clonality:
    Monoclonal
  • Clone:
    273210
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    3685
  • Antigen Synonyms:
    MSK8|Vitronectin Receptor Subunit Alpha|CD51|ITGAV|Vitronectin Receptor|VNRA|VTNR
  • Amino Acid Number:
    Phe31 to Val992
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    Chinese hamster ovary cell line CHO-derived recombinant human Integrin alphaV/CD51 Phe31-Val992; Accession # NP_002201
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220018-910
  • Supplier no.:
    MAB12191

Specifications

About this item

Type: Primary
Antigen: ITGAV
Clonality: Monoclonal
Clone: 273210
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human