Order Entry
United States
ContactUsLinkComponent
Anti-EPGN Rat Monoclonal Antibody [clone: 187240]
Anti-EPGN Rat Monoclonal Antibody [clone: 187240]
  220018-828
Anti-EPGN Rat Monoclonal Antibody [clone: 187240]
  220018-828
 :  MAB1127
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    epithelial mitogen
  • Antigen Symbol:
    EPGN
  • Clonality:
    Monoclonal
  • Clone:
    187240
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    500 µg
  • Environmentally Preferable:
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    71920
  • Antigen Synonyms:
    epigen|2310069M11Rik
  • Amino Acid Number:
    Leu53 to Ala103
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    E. coli -derived recombinant mouse Epigen Leu53-Ala103; Accession # Q924X1.1
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220018-828
  • Supplier no.:
    MAB1127

 

 

Type: Primary
Antigen: EPGN
Clonality: Monoclonal
Clone: 187240
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse