Order Entry
United States
ContactUsLinkComponent
Anti-6xHis Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AD1.1.1]
Anti-6xHis Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AD1.1.1]
  220018-182
Anti-6xHis Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AD1.1.1]
  220018-182
 :  IC050P
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    6X HIS Affinity Tag
  • Antigen Symbol:
    6xHis
  • Clonality:
    Monoclonal
  • Clone:
    AD1.1.10
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Multiple
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Antigen Synonyms:
    Histidine Epitope Tag|His Tag|Histidine Tag
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    His-tagged peptide
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220018-182
  • Supplier no.:
    IC050P

 

 

Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple