Order Entry
United States
ContactUsLinkComponent
Anti-TM4SF1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 877621]
Anti-TM4SF1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 877621]
Catalog # 102883-862
Supplier:  R&D Systems
Anti-TM4SF1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 877621]
Catalog # 102883-862
Supplier:  R&D Systems
Supplier Number:  FAB8164P
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Transmembrane 4 L Six Family Member 1
  • Antigen Symbol:
    TM4SF1
  • Clonality:
    Monoclonal
  • Clone:
    877621
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    4071
  • Antigen Synonyms:
    Membrane component chromosome 3 surface marker 1|T4S1_HUMAN|Transmembrane 4 L6 family member 1|TAAL6,Tm4sf1|M3S1|Tumor-associated antigen L6.
  • Amino Acid Number:
    Met1 to Cys202
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    NS0 mouse myeloma cell line transfected with human TM4SF1/L6 Met1-Cys202; Accession # P30408
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    102883-862
  • Supplier no.:
    FAB8164P

Specifications

About this item

Type: Primary
Antigen: TM4SF1
Clonality: Monoclonal
Clone: 877621
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human