Order Entry
United States
ContactUsLinkComponent
 
Anti-ITGA7 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 334908]
  102882-532
Anti-ITGA7 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 334908]
  102882-532
 :  FAB3518N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Integrin Alpha 7
  • Antigen Symbol:
    ITGA7
  • Clonality:
    Monoclonal
  • Clone:
    334908
  • Conjugation:
    Alexa Fluor® 700
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    16404
  • Antigen Synonyms:
    Integrin alpha-7 light chain|Integrin alpha-7|UNQ406/PRO768|Integrin alpha-7 70 kDa form|ITGA7
  • Amino Acid Number:
    Phe34 to Pro1036
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin  alpha7 Phe34-Pro1036; Accession # NP_032424
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    102882-532
  • Supplier no.:
    FAB3518N

 

 

Type: Primary
Antigen: ITGA7
Clonality: Monoclonal
Clone: 334908
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse