Order Entry
United States
ContactUsLinkComponent
Anti-IL21R Goat Polyclonal Antibody (Biotin)
Anti-IL21R Goat Polyclonal Antibody (Biotin)
Catalog # 220014-522
Supplier:  R&D Systems
Anti-IL21R Goat Polyclonal Antibody (Biotin)
Catalog # 220014-522
Supplier:  R&D Systems
Supplier Number:  BAF991

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Interleukin 21 Receptor
  • Antigen Symbol:
    IL21R
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
  • Gene ID:
    50615
  • Antigen Synonyms:
    CD360|IL-21 Receptor|Novel interleukin receptor|UNQ3121/PRO10273|Interleukin-21 receptor|IL-21 receptor|NILR
  • Amino Acid Number:
    Cys20 to Pro236
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-21 R Subunit Cys20-Pro236; Accession # Q9HBE5
  • Purification:
    Antigen Affinity-purified
  • Cat. No.:
    220014-522
  • Supplier no.:
    BAF991

Specifications

About this item

Type: Primary
Antigen: IL21R
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human