Order Entry
United States
ContactUsLinkComponent
Anti-PF4 Goat Polyclonal Antibody (Biotin)
Anti-PF4 Goat Polyclonal Antibody (Biotin)
Catalog # 220014-411
Supplier:  R&D Systems
Anti-PF4 Goat Polyclonal Antibody (Biotin)
Catalog # 220014-411
Supplier:  R&D Systems
Supplier Number:  BAF795

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Platelet Factor 4
  • Antigen Symbol:
    PF4
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
  • Gene ID:
    5196
  • Antigen Synonyms:
    SCYB4|PF-4|CXCL4
  • Amino Acid Number:
    Glu32 to Ser101
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    E. coli -derived recombinant human CXCL4/PF4 (R&D Systems, Catalog # 795-P4) Glu32-Ser101; Accession # P02776
  • Purification:
    Antigen Affinity-purified
  • Cat. No.:
    220014-411
  • Supplier no.:
    BAF795

Specifications

About this item

Type: Primary
Antigen: PF4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human