Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:S100 Calcium binding Protein A9
- Antigen Symbol:S100 A9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Gene ID:6280
- Antigen Synonyms:S100A9|S100 Calcium binding protein A9|CAGB|MRP14|CFAG|S100 calcium-binding protein A9|NIF|MAC387|MIF|L1AG|calgranulin B|60B8AG|CGLB|P14|S100 calcium-binding protein A9 (calgranulin B)|LIAG|S100 Calcium binding protein A9 (calgranulin B)
- Amino Acid Number:The2 to Pro114
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
- Immunogen:E. coli -derived recombinant human S100A9 The2-Pro114; Accession # P06702
- Purification:Antigen Affinity-purified
- Cat. No.:102869-710
- Supplier no.:AF5578
Specifications
About this item
Type: Primary
Antigen: S100 A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human