Order Entry
United States
ContactUsLinkComponent
Universal Spill Response Plus Roll, Dimpled and Perforated, Brady®
Universal Spill Response Plus Roll, Dimpled and Perforated, Brady®
Catalog # 10796-234
Supplier:  Brady Worldwide
undefined
Universal Spill Response Plus Roll, Dimpled and Perforated, Brady®
Catalog # 10796-234
Supplier:  Brady Worldwide
Supplier Number:  123450

Specifications

  • Description:
    Roll
  • Environmentally Preferable:
  • Class Type:
    Universal - All Purpose
  • Color:
    Gray
  • Dimensions:
    7.5W"x 50L'
  • Cat. No.:
    10796-234
  • Absorbency:
    3.3 gal.

Specifications

About this item

The SRP75P will absorb just about any liquid

This smaller absorbent is great for preventing slips, trips and falls that commonly occur in the retail, hospitality and grocery markets. It is designed to be highly absorbent, durable and reduce waste. Its small convenient size makes it ideal for employees to fold and carry in their pockets for quick response to spills.