Order Entry
United States
ContactUsLinkComponent
Universal Plus Spill Response Kit, 2 Gallon, Brady®
Universal Plus Spill Response Kit, 2 Gallon, Brady®
Catalog # 10796-470
Supplier:  Avantor
undefined
Universal Plus Spill Response Kit, 2 Gallon, Brady®
Catalog # 10796-470
Supplier:  Avantor

Specifications

  • Description:
    Portable Spill Kit
  • Environmentally Preferable:
  • Class Type:
    Chemical
  • Color:
    White
  • Dimensions:
    10.5H x 9.75" Dia.
  • Cat. No.:
    10796-470
  • Absorbency:
    4.5 gal.

Specifications

About this item

  • This 2-gallon universal absorbent spill kit is ideal for small spill responses
  • Kit includes 2 gallon container, 20 pads, 1 bag, tie, a pair of gloves, goggles, and instruction sheet