Condenser Box
Catalog # 470219-540
Supplier: Wards
Specifications
- Description:Condenser Box
- Cat. No.:470219-540
- Supplier no.:PH1136
Specifications
About this item
This condenser box provides different values of Electrolytic capacitors, ranging from 1μF to 4.7μF. Built on a 8 x 5" plastic box. All these capacitors are of ± 10% tolerance and are of 63 volts working voltage.