High Torque Generator
Catalog # 470219-358
Supplier: Vernier Software & Technology
Specifications
- Description:High Torque Generator 3 pack
- Cat. No.:470219-358
- Supplier no.:KW-HIGEN3
Specifications
About this item
The High Torque Generator is another option for your wind turbine generator. When compared to the standard Wind Turbine Generator, this High Torque Generator will produce lower voltage but higher current for a given rotor RPM. The High Torque Generator is great for load devices with higher current draw, such as the water pump. This generator has the same 2 mm driveshaft as the standard Wind Turbine Generator, so the Wind Turbine Hub is interchangeable with either motor. The generator also has 4-foot leads with shrink-wrap reinforced soldering.