Wind Turbine Generator
Catalog # 470219-356
Supplier: Vernier Software & Technology
Specifications
- Description:Wind Turbine Generator 10 Pack
- Cat. No.:470219-356
- Supplier no.:KW-GEN10
Specifications
About this item
This is the primary generator for wind turbine experiments because it runs smoothly and provides high power output at a relatively low RPM. This is the competition generator used in KidWind Challenge events. This generator has 4-foot leads which are attached with shrink-wrapped, reinforced soldering. Typical output in non-geared wind devices is 0.5–2.5 V at 0.1–0.3 A.