ContactUsLinkComponent
DuPont™ Tyvek® 400 FC Boot Covers with Skid Resistant Sole
Catalog # 10016-062
Supplier:  DuPont
undefined
DuPont™ Tyvek® 400 FC Boot Covers with Skid Resistant Sole
Catalog # 10016-062
Supplier:  DuPont
Supplier Number:  FC454SGY00010000
Specifications
  • Garment application:
    Laboratory
  • Color:
    Gray
  • Height:
    18"
  • Cat. No.:
    10016-062
  • Supplier no.:
    FC454SGY0000
Specifications
About this item
Tyvek® 400 FC protective fabric features a skid resistant friction coating.
  • Skid-resistant sole
  • Elastic top
  • Coverage up to calf
  • One size fits most
  • Serged seams
Used exclusively for shoe and boot covers and skid-resistant booties in select Tyvek® coveralls.
Certifications: Thickness meets ASTM D1777.