SP Bel-Art Bottle, Wide Mouth, with Screw Cap, Bel-Art Products, a part of SP
Catalog # 470219-166
Supplier: Bel-Art
Specifications
- Description:Bottle with Screw Cap, 237 mL
- Cat. No.:470219-166
- Supplier no.:106259006
Specifications
About this item
Wide mouth bottle, HDPE, cylindrical, screw closure.
- Size: 45 mm
- Capacity: 237 ml