Order Entry
United States
ContactUsLinkComponent
Windsoft® Nonperforated Roll Towels
Windsoft® Nonperforated Roll Towels
  102866-260
undefined
Windsoft® Nonperforated Roll Towels
  102866-260
 :  WIN108

 

  • Ø:
    5.5"
  • Width:
    8"
  • Cat. No.:
    102866-260
  • Roll length:
    350'
  • Supplier no.:
    WIN108
  • Ply:
    1

 

 

Roll towels are designed for hardwound roll towel dispensers.

  • Absorbent
  • Natural color
  • Paper material

Quality construction enables multipurpose versatility.