ContactUsLinkComponent
RNeasy® Plus Total RNA Extraction Kits, Qiagen
RNeasy® Plus Total RNA Extraction Kits, Qiagen
Catalog # 220006-476
Supplier:  Qiagen
CAS Number:  
RNeasy® Plus Total RNA Extraction Kits, Qiagen
Catalog # 220006-476
Supplier:  Qiagen
Supplier Number:  74134
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    RNeasy Plus Mini Kit
  • Cat. No.:
    220006-476
  • Includes:
    50 RNeasy Mini Spin Columns, gDNA Eliminator Spin Columns, Collection Tubes, RNase-Free Water and Buffers
  • Supplier no.:
    74134
Specifications
About this item
For phenol free total RNA extraction from cells/tissues with removal of genomic DNA contamination.
  • Efficient gDNA removal with unique gDNA Eliminator columns (no need for DNase)
  • High-quality total RNA in minutes using fast and simple extraction protocols
  • Phenol-free RNA isolation
  • Ideal for sensitive applications such as real-time RT-PCR and RNA-Seq
  • Sample size 30 mg
The RNeasy Plus Kits are next generation QIAGEN RNA extraction kits, combining high-quality RNA purification with effective on-column gDNA removal. Total RNA of high purity and yield is obtained from a wide range of cultured cells and easy-to-lyse tissues. The RNeasy Plus Mini Kit purifies up to 100 µg total RNA (>200 nt) from a single extraction with efficient gDNA removal. The RNeasy Plus Micro Kit is specially designed for limited amounts of samples and isolates up to 45 µg pure total RNA. The kits can be automated on the QIAcube and samples can be conveniently stabilized using RNAlater RNA Stabilization Reagent or Allprotect Tissue Reagent.