ContactUsLinkComponent
Wide Bite Tischler Biopsy Punch Forcep, OR Grade, Sklar
Wide Bite Tischler Biopsy Punch Forcep, OR Grade, Sklar
Catalog # 10184-290
Supplier:  Sklar
undefined
Wide Bite Tischler Biopsy Punch Forcep, OR Grade, Sklar
Catalog # 10184-290
Supplier:  Sklar
Supplier Number:  90-9560
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Material:
    Stainless steel
  • Tip configuration:
    Elliptical / Oval Bite with 1 Lower Tooth
  • Cat. No.:
    10184-290
  • Tip style:
    Straight
  • Supplier no.:
    90-9560
Specifications
About this item
Wide Bite Tischler Biopsy Punch Forceps are an OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy.
The jaws of this biopsy punch are designed to open 60 degrees, which is wider than other Tischler punches, in order to obtain tissue samples from obscure sites. This product is straight with an 8" shaft and 3 x 7-mm oval bite. It features a single, lower tooth used to grasp difficult to reach tissue and a locking mechansim above the horn on the handle so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal.
This reduces the risk of losing the tissue sample as the instrument is withdrawn. This product has a plier style handle. The lower handle also has two finger rests and a hook for user comfort and increased control.