Order Entry
United States
ContactUsLinkComponent
Iowa Puncturing Forceps, OR Grade, Sklar
Iowa Puncturing Forceps, OR Grade, Sklar
  10184-236
 :  Sklar
undefined
Iowa Puncturing Forceps, OR Grade, Sklar
  10184-236
 :  Sklar
 :  91-4310

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Length:
    254 mm (10")
  • Material:
    Stainless steel
  • Tip configuration:
    5 x 6 Teeth
  • Cat. No.:
    10184-236
  • Tip style:
    Double / 'S' Curved Blades
  • Supplier no.:
    91-4310

 

 

Iowa Membrane Puncturing Forceps are an OB/GYN instrument used to puncture the membrane of the amniotic sac and release the amniotic fluid.

They are ratcheted, finger ring forceps with 5 x 6 teeth, that provide a secure hold on the membranes. They have long, narrow shanks and jaws that each have a slight curvature.