ContactUsLinkComponent
Sklarlite™ Littauer Stitch Scissors, OR Grade, Sklar
Sklarlite™ Littauer Stitch Scissors, OR Grade, Sklar
Catalog # 10194-532
Supplier:  Sklar
undefined
Sklarlite™ Littauer Stitch Scissors, OR Grade, Sklar
Catalog # 10194-532
Supplier:  Sklar
Supplier Number:  23-1190
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    140 mm (5½")
  • Material:
    Stainless steel
  • Cat. No.:
    10194-532
  • Supplier no.:
    23-1190
  • Blade type:
    Straight, Blunt/Blunt
Specifications
About this item
Sklarlite™ Littauer Stitch Scissors are used for suture removal.
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They have larger, less delicate tips than Spencer Stitch Scissors, and are used for removing larger sutures. They should not be used for cutting tissue.
Sklarlite™ instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.