- Sterility:Non sterile
- Description:Tischler Oval Biopsy Punch
- Material:Stainless steel
- Environmentally Preferable:
- Tip configuration:Elliptical / Oval Bite with 1 Lower Tooth
- Curvature:Straight
- Cat. No.:10193-958
- Supplier no.:90-7520
Sklar's® Tischler Elliptical / Oval Biopsy Punch is a non-ratcheted, double spring OB/GYN instrument used in colposcopy procedures to obtain cervical tissue samples for biopsy.
- 0.3 x0.7 cm Bite
- Oval shaped tissue sample
- Pistol style handle with one large ring for multiple fingers
- Censitrac Ready products are industry standard and constructed of stainless steel
- Reusable
- Latex-free
It features a single, lower tooth used to grasp difficult to reach tissue and a locking mechanism above the horn on the handle so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal
A standard size biopsy punch may produce cause more bleeding and pain than a mini or baby punch; however, with certain diagnoses it is necessary to obtain a larger tissue sample. The length of the cannula/tip is approximately 8".
This reduces the risk of losing the tissue sample as the instrument is withdrawn.