Staticide® Restorer & Cleaner, Anti-Static Floor Cleaner, ACL
Supplier: ACL Staticide
Certificates
About this item
ACL Staticide Restorer Cleaner is a specialized anti-static detergent for ESD floor finishes. The cleaner renews static decay properties, surface resistivity, and the durability of ACL floor finishes. Special conductive additives clean while simultaneously breaking down electrical discharge and returning floor finishes to safe specifications.
- Removes black heel marks
- Lengthens maintenance cycle
- Revitalizes gloss
- Dilution ratio 1:10 (1 part cleaner to 10 parts water)
- Covers approximately 1000 - 1500 square feet per diluted gallon
No harmful chemicals like ammonia, phosphates, alkali, nonylphenol (NP), or nonylphenol ethoxylate (NPE) are used in the formulation. This special detergent can be applied with a mop or in a standard burnishing machine for easy application.
Ideal for the following applications:
• Electronic manufacturing facilities
• Maintaining ESD Floor Finishes
• Maintaining ESD Floor Tiles
Frequently Bought Together