Order Entry
United States
ContactUsLinkComponent
Jorgenson Scissors, OR Grade, Sklar
Jorgenson Scissors, OR Grade, Sklar
Catalog # 10015-654
Supplier:  Sklar
undefined
Jorgenson Scissors, OR Grade, Sklar
Catalog # 10015-654
Supplier:  Sklar
Supplier Number:  52-2979

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Length:
    305 mm (12")
  • Material:
    Stainless steel
  • Cat. No.:
    10015-654
  • Supplier no.:
    52-2979
  • Blade type:
    Strongly Curved, Blunt/Blunt

Specifications

About this item

Jorgenson Scissors are industry standard.