About this item
Econo™ Sterile Baumgartner Needle Holder is used for holding suture needles.
It has shorter jaws than both Crile-Wood and Mayo-Hegar Needle Holders. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan.
Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo™ Sterile instruments significantly reduce the likelihood of cross-contamination.