Order Entry
United States
ContactUsLinkComponent
Schnidt Tonsil Forceps, OR Grade, Sklar
Schnidt Tonsil Forceps, OR Grade, Sklar
Catalog # 10193-724
Supplier:  Sklar
undefined
Schnidt Tonsil Forceps, OR Grade, Sklar
Catalog # 10193-724
Supplier:  Sklar
Supplier Number:  74-3075
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Schnidt Tonsil Forceps
  • Length:
    19 cm (7.5")
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Curvature:
    Half Curved
  • Cat. No.:
    10193-724
  • Tip style:
    Serrated
  • Grade:
    Premium OR-Grade
  • Sterile:
    -
  • Supplier no.:
    74-3075

Specifications

About this item

Sklar's® Schnidt Tonsil Forceps are used for placing packing following tonsil removal.

This instrument with a partial curve are also known as Schnidt-Boettcher forceps

Censitrac Ready products are industry standard and constructed of stainless steel.

If the instrument has a full curve, it is also known as Schnidt-Sawtell forceps. This product has a slight curve and one open finger ring.

Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.