ContactUsLinkComponent
Tischler Micro-Cut Oval Biopsy Punch, OR Grade, Sklar
Tischler Micro-Cut Oval Biopsy Punch, OR Grade, Sklar
Catalog # 10183-848
Supplier:  Sklar
undefined
Tischler Micro-Cut Oval Biopsy Punch, OR Grade, Sklar
Catalog # 10183-848
Supplier:  Sklar
Supplier Number:  90-8940
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Sterility:
    Non sterile
  • Description:
    Micro-Cut Tischler Oval Biopsy Punch with Locking Tip
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Tip configuration:
    Elliptical/Oval Bite; 1 Lower Tooth
  • Curvature:
    Straight
  • Cat. No.:
    10183-848
  • Supplier no.:
    90-8940
Specifications
About this item
Sklar's® Micro-Cut Tischler Biopsy Punch Forceps are an OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy.
  • 0.3 x 0.7 cm Bite
  • Oval shaped tissue sample
  • Pistol style handle with one large ring for multiple fingers
  • Reusable
  • Latex-free
It features a single, lower tooth used to grasp difficult to reach tissue and a locking mechanism above the horn on the handle so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal
Censitrac Ready OR Grade Micro-Cut Tischler Oval Biopsy Punch is industry standard and constructed of stainless steel.
This reduces the risk of losing the tissue sample as the instrument is withdrawn. A standard size biopsy punch may produce greater bleeding and pain than a mini or baby punch.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.