ContactUsLinkComponent
Sklar Blue™ Potts-Smith Tissue Forceps, OR Grade, Sklar
Sklar Blue™ Potts-Smith Tissue Forceps, OR Grade, Sklar
Catalog # 10194-078
Supplier:  Sklar
undefined
Sklar Blue™ Potts-Smith Tissue Forceps, OR Grade, Sklar
Catalog # 10194-078
Supplier:  Sklar
Supplier Number:  91-5413
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    210 mm (8¼")
  • Material:
    Stainless Steel with Non-Conductive Nylon Coating
  • Environmentally Preferable:
  • Tip configuration:
    1 x 2 Teeth
  • Cat. No.:
    10194-078
  • Tip style:
    Straight
  • Supplier no.:
    91-5413
Specifications
About this item
Blue™ Potts-Smith Tissue Forceps are used for grasping fine or delicate tissue in electrosurgical procedures
Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue.
Sklar Blue™ instruments are autoclavable and may be cleaned ultrasonically.
These narrow forceps grasp the tissue, but also have a stop peg to prevent a harsh grasp that may crush the tissue.
Certifications: All Sklar Blue™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Blue™ electrosurgical coatings are guaranteed for one year against manufacturing defects.