ContactUsLinkComponent
Precision Suture Scissors, OR Grade, Sklar
Precision Suture Scissors, OR Grade, Sklar
Catalog # 89025-316
Supplier:  Sklar
undefined
Precision Suture Scissors, OR Grade, Sklar
Catalog # 89025-316
Supplier:  Sklar
Supplier Number:  22-2822
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    89 mm (3½")
  • Material:
    Stainless steel
  • Cat. No.:
    89025-316
  • Supplier no.:
    22-2822
  • Blade type:
    Curved, Sharp/Sharp
Specifications
About this item
Precision Suture Scissors are used for suture removal
This instrument has sharp tips, that easily slide under and gently lift sutures in preparation for removal. One of the sharp/sharp blades has a notch that the suture rests in before it is cut.