About this item
PP, translucent.
- Able to tolerate centrifuge speeds up to 20000 RCF
- Suitable for IP and mass spectrometry applications
- Certified RNase/DNase-free
Thermo Scientific™ Pierce™ Low protein binding microcentrifuge tubes (1.5 ml) help minimize protein sample loss during sample manipulation and are ideal in applications where processing steps are extensive.