Order Entry
United States
ContactUsLinkComponent
Pierce™ Immobilized Anti-TMT™ Antibody Resin, Thermo Scientific
Pierce™ Immobilized Anti-TMT™ Antibody Resin, Thermo Scientific
Catalog # PI90076
Supplier:  Invitrogen
CAS Number:  
Pierce™ Immobilized Anti-TMT™ Antibody Resin, Thermo Scientific
Catalog # PI90076
Supplier:  Invitrogen
Supplier Number:  90076
CAS Number:  

Specifications

  • Description:
    Immobilized Anti-TMT™ antibody resin
  • Size:
    6 mL
  • Storage Conditions:
    Upon receipt store all products at 4˚C.
  • Shipping Temperature:
    Product shipped with an ice pack.
  • Cat. No.:
    PI90076
  • Supplier no.:
    90076

Specifications

About this item

Anti-TMT™ Antibody resin enables selective enrichment of peptides in complex samples that have been labeled with Tandem Mass Tag™ Reagents.

  • Specific — uses specific, high-affinity antibody for the TMT structure (Kd>100pM)
  • Compatible — effective for all varieties of TMT Reagents (amine-reactive or cysteine-reactive)
  • Versatile — use complete kit for sample enrichment to simplify analysis, or choose the standalone resin or standalone antibody to develop your own purification and detection strategies

Tandem Mass Tag™ (TMT™) Reagents are used to multiplex-label and measure proteins in different samples using tandem mass spectrometry. This resin is agarose-immobilized anti-TMT monoclonal antibody, which is effective to capture and affinity purify peptides or proteins that have been labeled using any TMT Reagent. The strategy is effective for simplifying otherwise complex samples before running the mass analysis, especially when the targets for labeling are rare (e.g., cysteine sulfhydryls). Enrichment of labeled peptides by this method results in fewer peptides identified per protein but can significantly increase the number of unique proteins identified, making it easier to detect and measure low-abundance proteins.

Caution: For Research Use Only. Not for use in diagnostic procedures.