Specifications
- Description:Phosphoprotein Enrichment Kit, 10 columns
- Size:1 mL
- Storage Conditions:Upon receipt store at 4˚C.
- Cat. No.:PI90003
- Supplier no.:90003
Specifications
About this item
Phosphoprotein Enrichment Kit is designed for the efficient enrichment of phosphorylated proteins derived from mammalian cells and tissues.
- Specific – low nonspecific protein contamination from complex biological samples, such as cell culture lysate and mouse tissue extract
- Fast – easy spin format enables enrichment of phosphorylated proteins in less than 2 hours
- Superior yield – acheives higher yields than other commercially available kits
- Convenient format – complete kit offering pre-dispensed spin columns, buffers, reagents and ultrafiltration columns for enrichment
- Compatible – downstream applications include mass spectrometry, Western blotting, and 2D-PAGE
The proprietary resin and buffer composition produces superior yields with negligible non-specific binding. The kit is uses a convenient spin column format and is compatible with downstream applications including Western blotting, Mass Spec, 2D-PAGE, and protein arrays. The affinity resin is also available as pre-packed chromatography cartridges for use with automated liquid chromatography (LC) systems.