Order Entry
United States
ContactUsLinkComponent
Pierce™ Mem-PER™ Plus Membrane Protein Extraction Kit, Thermo Scientific
Pierce™ Mem-PER™ Plus Membrane Protein Extraction Kit, Thermo Scientific
Catalog # PI89842
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Mem-PER™ Plus Membrane Protein Extraction Kit, Thermo Scientific
Catalog # PI89842
Supplier:  Invitrogen
Supplier Number:  89842
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Mem-PER™ Plus Membrane Protein Extraction Kit
  • Size:
    300 mL
  • Cat. No.:
    PI89842
  • Supplier no.:
    89842

Specifications

About this item

The Mem-PER™ Plus Membrane Protein Extraction Kit enables small-scale solubilization and enrichment of integral membrane proteins and membrane-associated proteins in a simple reagent-based procedure. It effectively isolates membrane proteins from cultured cells and tissues using a simple benchtop microcentrifuge procedure.

  • Extraction and isolation – produces minimal cross-contamination (typically less than 10%) of cytosolic protein into the membrane protein fraction
  • Cells or tissues – effective for extraction from cultured mammalian cells and mammalian tissues
  • Downstream compatibility – analyze membrane protein extracts by SDS-PAGE, Western blotting, immunoprecipitation and protein assays
  • Fast and simple – isolation of membrane proteins in approximately one hour
  • No special equipment required – only a benchtop microcentrifuge, tubes, homogenizers, and pipettors are needed

The Mem-PER Plus Kit effectively isolates membrane proteins from cultured cells and tissues using a simple benchtop microcentrifuge procedure

Store Cell Wash Solution and Solubilization Buffer at 4˚C. Aliquot and store Permeabilization Buffer at -20˚C to avoid freeze/thaws. Product is shipped on ice.

Integral membrane proteins having one or two membrane-spanning domains are effectively solubilized and isolated from cytosolic proteins. The kit makes possible the extraction and selective enrichment of integral and attached membrane proteins from cultured mammalian cells, as well as from hard and soft mammalian tissues. The Mem-PER Plus Kit has many advantages compared to our original Mem-PER Kit. The protocol is simpler, and the fractions are directly compatible with many downstream applications such as SDS-PAGE, Western Blotting, BCA, immunoprecipitation, and amine-reactive protein labeling techniques.

Delivery: The kit contains a cell wash solution (225 mL), permeabilization buffer (50 mL), and solubilization buffer (25 mL).
Caution: For Research Use Only. Not for use in diagnostic procedures.