Order Entry
United States
ContactUsLinkComponent
Pierce™ Reserpine Standard for LC-MS, Thermo Scientific
Pierce™ Reserpine Standard for LC-MS, Thermo Scientific
Catalog # PI88326
Supplier:  Invitrogen
CAS Number:  
Pierce™ Reserpine Standard for LC-MS, Thermo Scientific
Catalog # PI88326
Supplier:  Invitrogen
Supplier Number:  88326
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Pierce™ Reserpine Standard for LC-MS, Sufficient for 5 to 500 Injections
  • Size:
    5×1 ml
  • Cat. No.:
    PI88326
  • Contents:
    100 pg/uL reserpine in 50% isopropanol
  • Supplier no.:
    88326

Specifications

About this item

Pierce Reserpine Standard for LC-MS is a precise concentration of reserpine specifically designed for performance evaluation of mass spectrometers, including the Thermo Scientific ion trap and TSQ series of instruments.

  • Pre-Diluted and Requires Minimal Additional Dilutions to Reach Target Concentration for Injection
  • Easily Removed and Replaced When Withdrawing Solution
  • Mass Spectrometry Grade Reagent in a Non-Leachable Screw-Cap Vials
  • Store at 4˚C for up to One Year

The standard is provided in a concentration of 100pg/µL in 50% isopropyl alcohol and requires minimal additional dilutions. The product is provided in a pack of 5 x 1 mL glass amber vials with PTFE-lined screw caps. Pierce Reserpine Standard for LC-MS is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications.

Caution: For Research Use Only. Not for use in diagnostic procedures.