About this item
Zeba™ 96-well Spin Desalting Plates (40K MWCO) contain a size-exclusion chromatography resin that provides excellent protein desalting performance in a centrifuge format for 5 µL to 4 mL samples.
- High recovery, low-binding resin maximizes protein recovery
- No screening of fractions or waiting for protein to emerge by gravity-flow
- Excellent recovery for protein larger than 40,000 Da
Zeba™ Spin Desalting Plates are polypropylene devices containing a proprietary high-performance size-exclusion resin for effective removal of salts and other small molecules (<2000 daltons) while providing excellent recovery for protein larger than 40,000 daltons
The Zeba™ 96-well Spin Desalting Plates require no resin dispensing or hydration and provide the same high protein recovery as spin columns with at least 95% removal of salts and other small molecules. One plate can process 96 small sample volumes (20 to 100 µL) in 5 minutes.
Zeba™ Spin Desalting Plates provide trouble-free desalting and buffer exchange for sample volumes ranging from 5 µL to 4 mL. This 96-well spin plate format enables more high-throughput processing.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot NumberThermo_Scientific_Complete_Protein_Clean-Up_Toolbox_Brochure