To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Pierce™ Digestion Indicator for Mass Spectrometry
- Size:10 µg
- Environmentally Preferable:
- Storage Conditions:store at -20˚C. Product is shipped on dry ice.
- Cat. No.:PI84841
- Supplier no.:84841
Specifications
About this item
The Digestion Indicator for Mass Spectrometry is a non-mammalian recombinant protein (26kDa) with five signature peptides for use in determining the digestion efficiency and reproducibility across multiple samples.
- Versatile
- For use with mass spectrometer
- Non-mammalian protein source
This product is sufficient for production of five signature peptides upon digestion for mass spectrometry.