Order Entry
United States
ContactUsLinkComponent
Pierce Subcellular Protein Fractionation Kit, Thermo Scientific
Pierce Subcellular Protein Fractionation Kit, Thermo Scientific
Catalog # PI78840
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce Subcellular Protein Fractionation Kit, Thermo Scientific
Catalog # PI78840
Supplier:  Invitrogen
Supplier Number:  78840
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    35 mL
  • Cat. No.:
    PI78840
  • Supplier no.:
    78840

Specifications

About this item

Thermo Scientific Subcellular Protein Fractionation Kit for Cultured Cells enables segregation and enrichment of proteins from five different cellular compartments.

  • Concise—extract functional cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal protein fractions from mammalian cultured cells in less than 3 hours
  • Convenient—perform a simple procedure without using ultracentrifugation over gradients
  • Complete—obtain cytoplasmic, membrane, soluble nuclear, chromatin-bound and cytoskeletal protein fractions from a single kit
  • Cultured Cells—developed and optimized for use with cultured mammalian cells

Fractionation of subcellular proteins enables protein localization assessment and protein enrichment from specific cellular compartments. Thermo Scientific Subcellular Protein Fractionation Kit includes a combination of reagents for stepwise lysis of cells into functional cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal protein fractions in less than 3 hours. Extracts from each subcellular compartment generally have less than 15% contamination between fractions, which is sufficient purity for most experiments studying protein localization and redistribution.

Ordering information: Upon receipt store kit at -20 °C, or store individual components as indicated above. Kit is shipped with dry ice.
Caution: For Research Use Only. Not for use in diagnostic procedures.