Pierce™ North2South™ Hybridization Stringency Wash Buffer (2X), Thermo Scientific
Catalog # PI-37555
Supplier: Invitrogen
CAS Number:
Specifications
- Description:North2South™ hybridization stringency wash buffer (2X)
- Size:375 mL
- Cat. No.:PI-37555
- Supplier no.:37555
Specifications
About this item
Thermo Scientific North2South Hybridization Stringency Wash Buffer (2X), for use with the North2South Chemiluminescent Hybridization and Detection Kit.
- pH 6.9
- Aqueous buffer
Caution: For Research Use Only. Not for use in diagnostic procedures. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8˚C.