ContactUsLinkComponent
Pierce™ NBT Substrate Powder (nitro-blue tetrazolium chloride), Thermo Scientific
Pierce™ NBT Substrate Powder (nitro-blue tetrazolium chloride), Thermo Scientific
Catalog # PI34035
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ NBT Substrate Powder (nitro-blue tetrazolium chloride), Thermo Scientific
Catalog # PI34035
Supplier:  Invitrogen
Supplier Number:  34035
CAS Number:  
Specifications
  • Description:
    NBT Powder
  • Size:
    1 g
  • Cat. No.:
    PI34035
  • Supplier no.:
    34035
Specifications
About this item
NBT (nitro-blue tetrazolium chloride) yields a blue-purple, water-insoluble product upon reduction of the tetrazolium salt
  • Upon receipt store product at 4˚C protected from moisture
  • Product is shipped at ambient temperature.
The product of reduction is NBT-formazan. This substrate is widely used for immunochemistry applications because the color is linear and stable. NBT is used as the substrate for both glucose oxidase and alkaline phosphatase systems
Caution: For Research Use Only. Not for use in diagnostic procedures.