Pierce™ EZ-Link™ Maleimide Activated Horseradish Peroxidase Kit, Thermo Scientific
Supplier: Invitrogen
Certificates
About this item
Thermo Scientific Pierce Maleimide Activated Horseradish Peroxidase is for preparation of HRP conjugates with proteins, peptides or other ligands that contain sulfhydryl groups, such as reduced cysteines.
- Activated HRP
- Sulfhydryl-reactive
- Product is shipped at ambient temperature
This kit contains 5 mg of conjugated protein and can effectively modify 5 mg of immunoglobulin and also contains all buffers and one 10 mL desalting column.
Total weight is
25mg as a result of buffer components and salts lyophilized along with the activated protein.
Delivery: Kit contains two modification reagents allowing flexibility in the method of producing free sulfhydryls on macromolecules. 2-MercaptoethylamineHCl is a mild reagent for reduction of IgG and F(ab´)2 fragments. SATA is a sulfhydryl-containing modification reagent that reacts with primary amines to present protected sulfhydryl groups, which can be exposed upon treatment with hydroxylamine.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Specifications
- Storage Conditions:4˚C