Order Entry
United States
ContactUsLinkComponent
Pierce™ Immunodiffusion Plates, Multiple Pattern, Thermo Scientific
Pierce™ Immunodiffusion Plates, Multiple Pattern, Thermo Scientific
Catalog # PI31111
Supplier:  Invitrogen
Pierce™ Immunodiffusion Plates, Multiple Pattern, Thermo Scientific
Catalog # PI31111
Supplier:  Invitrogen
Supplier Number:  31111

Specifications

  • Size:
    6 plates
  • Cat. No.:
    PI31111
  • Supplier no.:
    31111

Specifications

About this item

These Immunodiffusion Plates are standard Ouchterlony gel plates containing patterns of six wells around one well for studying and characterizing antibody-antigen binding interactions.

  • Gelling agent contains precipitin brighteners for easy visualization
  • Contains diffusion enhancers to help speed the interaction process
  • Excellent precipitin bands with antibodies from all species (including rabbit antibodies)
  • Gels can be washed, dried and stained for a permanent record

Immunodiffusion (ID) is a classic technique for detecting antibody-antigen interactions based on the theory of double diffusion originally described by Oudin and Ouchterlony

When multivalent antigens combine with divalent antibodies in solution, three-dimensional lattices are formed that aggregate and precipitate. The amount of precipitate varies in proportion to the concentration of the antigen and antibody. At equivalent or optimal proportions almost all the antigen and antibody will precipitate. If there is an excess of antibody, the complexes formed with the antigen are insoluble. When there is an excess of antigen, the precipitate has a tendency to dissolve caused by the formation of soluble complexes.

Antigens and antibodies are placed into separate wells that are cut into a gel matrix and allowed to diffuse towards each other. If the reaction is positive, a precipitate forms that appears as an opaque line. The precipitation reaction occurs when the antigen and antibody concentrations are combined at near equivalent proportions.

Storage conditions: Upon receipt store at 4 °C. Do not freeze plates or allow them to become dry.

Caution: For Research Use Only. Not for use in diagnostic procedures.